Environmental Express® Disposable Distillation Micro Tubes
Save time and improve safety in your lab
  • Eliminate broken glassware—made of polypropylene with hydrophobic membrane and cap
  • No more having to wash glass tubes—just use and toss!
  • Molded-in graduations at 6- and 10-mL for easy measuring
  • Score-line for quick separation of collection tube
  • Compatible with most micro digestion systems
Micro tubes scale down sample and reagent volumes to less than 10 mL for rapid distillations of cyanide, phenols, and ammonia. These user-filled polypropylene tubes work with most micro distillation (Lachat) systems.
The polypropylene tubes consist of two key pieces with a hydrophobic membrane. Choose a tube for cyanide, ammonia, or phenol. Tubes have one press-fit joint that will never leak—and are simply disposed of after use.
7,100.00 - ₹198,000.00INR / Pkg of 100
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Disposible Distillation Micro Tubes, Cyanide; 10/Pk
₹23,600.00INR / Pkg of 10
COMPARE
Disposible Distillation Micro Tubes, Cyanide; 21/Pk
₹48,800.00INR / Pkg of 21
COMPARE
Disposible Distillation Micro Tubes, Cyanide; 26/Pk
₹59,400.00INR / Pkg of 26
COMPARE
Disposible Distillation Micro Tubes, Cyanide; 100/Pk
₹198,000.00INR / Pkg of 100
COMPARE
Disposible Distillation Micro Tubes, Phenol; 10/Pk
₹23,600.00INR / Pkg of 10
COMPARE
Disposible Distillation Micro Tubes, Phenol; 21/Pk
₹48,800.00INR / Pkg of 21
COMPARE
Disposible Distillation Micro Tubes, Phenol; 26/Pk
₹59,400.00INR / Pkg of 26
COMPARE
Disposible Distillation Micro Tubes, Phenol; 100/Pk
₹198,000.00INR / Pkg of 100
COMPARE
Disposable Measure Plug; 50/Pk
₹7,100.00INR / Pkg of 50
COMPARE
Replacement Top Cap; 50/Pk
₹7,500.00INR / Pkg of 50
COMPARE
Disposible Distillation Micro Tubes, Ammonia; 10/Pk
₹23,600.00INR / Pkg of 10
COMPARE
Disposible Distillation Micro Tubes, Ammonia; 21/Pk
₹48,800.00INR / Pkg of 21
COMPARE
Disposible Distillation Micro Tubes, Ammonia; 26/Pk
₹59,400.00INR / Pkg of 26
COMPARE
Disposible Distillation Micro Tubes, Ammonia; 100/Pk
₹198,000.00INR / Pkg of 100
Showing 1- 14 of 14