Environmental Express SimpleDist Micro C8251 Replacement Top Cap; 50/Pk
Representative image only.
Environmental Express SimpleDist Micro C8251 Replacement Top Cap; 50/Pk
Environmental Express | Item: 3520289
Save time and improve safety in your lab
₹7,500.00INR / Pkg of 50
₹7,500.00INR / Pkg of 50
Buy It Together
Current Item
Environmental Express SimpleDist Micro C8251 Replacement Top Cap; 50/Pk
Environmental Express SimpleDist Micro C8251 Replacement Top Cap; 50/Pk
3520289
₹7,500.00INR / Pkg of 50
+ =
Total Price 7500.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
tube assemblies with hydrophobic membranes, sample collection tubes, and caps.
Key Features
  • Eliminate broken glassware—made of polypropylene with hydrophobic membrane and cap
  • No more having to wash glass tubes—just use and toss!
  • Molded-in graduations at 6- and 10-mL for easy measuring
  • Score-line for quick separation of collection tube
  • Compatible with most micro digestion systems
More About this Item
Micro tubes scale down sample and reagent volumes to less than 10 mL for rapid distillations of cyanide, phenols, and ammonia. These user-filled polypropylene tubes work with most micro distillation (Lachat) systems.
The polypropylene tubes consist of two key pieces with a hydrophobic membrane. Choose a tube for cyanide, ammonia, or phenol. Tubes have one press-fit joint that will never leak—and are simply disposed of after use.
Specifications
  • DescriptionReplacement Top Cap; 50/Pk