Environmental Express C8021C Disposable Distillation Micro Tubes, Cyanide; 21/Pk
Representative image only.
Environmental Express C8021C Disposable Distillation Micro Tubes, Cyanide; 21/Pk
Environmental Express | Item: 3520271
Save time and improve safety in your lab
₹48,800.00INR / Pkg of 21
₹48,800.00INR / Pkg of 21
Buy It Together
Current Item
Environmental Express C8021C Disposable Distillation Micro Tubes, Cyanide; 21/Pk
Environmental Express C8021C Disposable Distillation Micro Tubes, Cyanide; 21/Pk
3520271
₹48,800.00INR / Pkg of 21
+ + =
Total Price 82000.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Includes
tube assemblies with hydrophobic membranes, sample collection tubes, and caps.
Key Features
  • Eliminate broken glassware—made of polypropylene with hydrophobic membrane and cap
  • No more having to wash glass tubes—just use and toss!
  • Molded-in graduations at 6- and 10-mL for easy measuring
  • Score-line for quick separation of collection tube
  • Compatible with most micro digestion systems
  • 3 month warranty
More About this Item
Micro tubes scale down sample and reagent volumes to less than 10 mL for rapid distillations of cyanide, phenols, and ammonia. These user-filled polypropylene tubes work with most micro distillation (Lachat) systems.
The polypropylene tubes consist of two key pieces with a hydrophobic membrane. Choose a tube for cyanide, ammonia, or phenol. Tubes have one press-fit joint that will never leak—and are simply disposed of after use.
3 Month Warranty
Specifications
  • Capacity (mL)10
  • DescriptionDisposible Distillation Micro Tubes, Cyanide; 21/Pk