DWK Kimble® Babcock Pipettes
Get accurate results, easily
  • Conforms to AOAC Method 989.04
  • Features permanent brown-stain scales
Model 34590-50 is designed to comply with the Pennsylvania modified Babcock as referenced in APHA 15.8. Models 34590-55,-56,-60, and -61 include a special rubber washer that allows air displaced by incoming milk to escape freely. Models 34590-55 and -56 meet ASTM Specification E1043, Type II A requirements; models 34590-60 and -61 meet ASTM Specification E1043, Type II B requirements. All pipettes feature permanent brown-stain scales.
5,100.00 - ₹56,800.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy (%) Max Volume (mL) Sterile
Availability Pricing
DWK Life Sciences (Kimble) Babcock Pipettes; skim milk test, to contain WW-34590-50 Mfr # 580S-9
COMPARE
±0.050mL 9 No
₹5,700.00INR / Each
DWK Life Sciences (Kimble) Babcock pipettes; milk test, to contain; 12/cs WW-34590-55 Mfr # 3005S-176
COMPARE
±0.050mL 17.6 No
₹56,800.00INR / Case of 12
DWK Life Sciences (Kimble) Glass Babcock Pipette, Milk Test, Calibrated "To Deliver" 17.6 mL; 1/Pack WW-34590-61 Mfr # 3001-176
COMPARE
±0.050mL 17.6 No
₹5,100.00INR / Each