Dual-Containment Tanks for Chemical Feed Systems
Exceeds EPA secondary containment requirements (49 CFR)
These chemical-resistant polypropylene tanks feature heavy-duty wall construction for liquids with specific gravity of 1.5. Tank tops come equipped with an integral pump mounting platform rated to 240 lb (113 kg) — easily mount heavy metering pumps or mixers directly on to the tank lid. Tanks come standard with an 8" vented twist-on lid, 1" or 2" NPT(F) spinwelds, plus a pump suction connection for 3/8" OD tubing, or 1/2" OD tubing when grommet is removed.
179,000.00 - ₹798,000.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (in) Material Volume (Gal)
Availability Pricing
Gemini 01-28907 Dual Containment Tank for Chemical Feed Systems, 15 Gallons; Blue 0693020 Mfr # 01-28907
COMPARE
27 Polypropylene 15
₹179,000.00INR / Each
Gemini 01-14871 Dual Containment Tank for Chemical Feed Systems, 20 Gallons; Blue 0693022 Mfr # 01-14871
COMPARE
25 1/2 Polypropylene 20
₹192,000.00INR / Each
Gemini 01-14874 Dual Containment Tank for Chemical Feed Systems, 40 Gallons; Blue 0693024 Mfr # 01-29436
COMPARE
38 1/2 Polypropylene 40
₹273,000.00INR / Each
Gemini 01-14877 Dual Containment Tank for Chemical Feed Systems, 62 Gallons; Blue 0693026 Mfr # 01-14877
COMPARE
37 Polypropylene 62
₹317,000.00INR / Each
Gemini 01-14870 Dual Containment Tank for Chemical Feed Systems, 120 Gallons; Blue 0693028 Mfr # 01-14870
COMPARE
46 Polypropylene 120
₹492,000.00INR / Each
Gemini 01-1073 Dual Containment Tank for Chemical Feed Systems, 20 Gallons; Natural 0693038 Mfr # 01-1073
COMPARE
25 1/2 Polypropylene 20
₹192,000.00INR / Each
Gemini 01-1076 Dual Containment Tank for Chemical Feed Systems, 40 Gallons; Natural 0693040 Mfr # 01-1076
COMPARE
38 1/2 Polypropylene 40
₹273,000.00INR / Each
Gemini 01-1074 Dual Containment Tank for Chemical Feed Systems, 220 Gallons; Natural 0693046 Mfr # 01-1074
COMPARE
48 Polypropylene 220
₹798,000.00INR / Each
Showing 1- 8 of 8