Gemini 01-14877 Dual Containment Tank for Chemical Feed Systems, 62 Gallons; Blue
Representative image only.
Gemini 01-14877 Dual Containment Tank for Chemical Feed Systems, 62 Gallons; Blue
Gemini | Mfr: 01-14877| Item: 0693026
Exceeds EPA secondary containment requirements (49 CFR)
₹317,000.00INR / Each
₹317,000.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
These chemical-resistant polypropylene tanks feature heavy-duty wall construction for liquids with specific gravity of 1.5. Tank tops come equipped with an integral pump mounting platform rated to 240 lb (113 kg) — easily mount heavy metering pumps or mixers directly on to the tank lid. Tanks come standard with an 8" vented twist-on lid, 1" or 2" NPT(F) spinwelds, plus a pump suction connection for 3/8" OD tubing, or 1/2" OD tubing when grommet is removed.
1 Year Warranty
Specifications
  • MaterialPolypropylene
  • Volume (Gal)62
  • Volume (Liters)235
  • Height (in)37
  • Height (cm)93.98
  • Outside Diameter (OD) (in)25 1/2
  • Outlet Connections2" NPT(F)
  • DescriptionDual Containment Tank for Chemical Feed Systems, 62 Gallons; Blue