Digi-Sense Thermocouple Economical Hollow Tube Probes
Hollow Tube probes are designed to be more compactat and at a lower cost
  • Response Time: 2-4 seconds or greater
  • Each probe comes with a color coded ANSI mini connector: type J-black, type K-yellow, type T-blue, and type E-purple.
  • Custom designs or changes to this product are available upon request
Note the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220°F (104°C); PVC handles are rated to 200°F (93°C); Stainless Steel handles are rated at 450°F (232°C); Mini Connectors are rated to 392°F (200°C).
9,100.00 - ₹10,800.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Junction Max Temperature (° F) Probe Type
Availability Pricing
COMPARE
Grounded 425 T
₹9,100.00INR / Each
COMPARE
Grounded 425 K
₹9,700.00INR / Each
COMPARE
Grounded 425 T
₹9,900.00INR / Each
COMPARE
Ungrounded 425 J
₹9,600.00INR / Each
COMPARE
Ungrounded 425 T
₹10,300.00INR / Each
COMPARE
Grounded 425 K
₹10,300.00INR / Each
COMPARE
Ungrounded 425 T
₹10,600.00INR / Each
COMPARE
Grounded 425 J
₹10,800.00INR / Each
Showing 1- 8 of 8