Digi-Sense® High-Temperature 25" Thermocouple Wire Probes with Stripped Leads
High-temperature thermocouple probes designed for multiple bends
  • Bend probes around pipe diameters as small as twice the sheath diameter to easily determine your pipe temperature.
  • Response Time: 1.5 seconds or greater
  • Type K and J feature ICONEL 600 sheath
  • Type T 316 Stainless Steel sheath
  • Probes are insulated with tightly packed powdered MgO in 25" long sheaths
  • Probes include 3-ft stranded 24 gauge (0.020" dia) fiberglass wire with stainless steel over braid.
  • Each probe comes with color coded ANSI miniconnectors: type J-black, type K-yellow, type T-blue, and type E-purple
  • Custom designs or changes to this product are available upon request.
Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220°F (104°C); PVC handles are rated to 200°F (93°C); Stainless Steel handles are rated at 450°F (232°C); Mini Connectors are rated to 392°F (200°C).
10,300.00 - ₹10,500.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Description
Availability Pricing
COMPARE
1690 Type-K, High-Temperature Wire Probe, 25" Length, 0.063" Diameter, Stripped Ends, Grounded, 3 ft 24-Gauge Cable
₹10,500.00INR / Each
COMPARE
500 Type-T, High-Temperature Wire Probe, 25" Length, 0.063" Diameter, Stripped Ends, Grounded, 3 ft 24-Gauge Cable
₹10,300.00INR / Each