Digi-Sense® Blue Spirit Certified Glass Thermometers, Celsius-scale
NIST-Traceable individually calibrated thermometers feature precise and accurate temperature measurement
  • Non-mercury blue spirit liquid
  • NIST-certified detailed, laminated report of calibration included
These thermometers are ideal for use in applications where a very precise accurate temperature measurement is required. Thermometers are individually calibrated to NIST standards at specific temperatures and supplied with an NIST report of calibration stating corrections to one-tenth the smallest division.
Thermometers feature triangular anti-roll fitting, permanently fused markings, blue spirit liquid, white background glass; are also individually serial numbered. Include an NIST-traceable statement of accuracy and a triangular-style antiroll fitting.
56,000.00 - ₹115,000.00INR / Each
17 variations of this product are available.
See All Product Options Below
17 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Points Min Temperature (° C) Description
Availability Pricing
COMPARE
-78, -20, 0, 4, 37°C -100 Calibrated Liquid-In-Glass Thermometer; -100 to 50C, 76mm Immersion, Organic Liquid Fill
Call For Price
COMPARE
0, 50, 100°C -20 Calibrated Liquid-In-Glass Thermometer, -20 to 110°C, 76 mm Immersion, Organic Liquid Fill, Calibration Points of 0, 50, 100°C
Call For Price
COMPARE
0, 60, 120°C -20 Calibrated Liquid-In-Glass Thermometer, -20 to 150°C, 76 mm Immersion, Organic Liquid Fill
Call For Price
COMPARE
0, 37, 56°C -10 Calibrated Liquid-In-Glass Thermometer, -10 to 70°C, 76 mm Immersion, Organic Liquid Fill
Call For Price
COMPARE
0, 37, 56°C 1 Calibrated Liquid-In-Glass Thermometer, 1 to 61°C, 76 mm Immersion, Organic Liquid Fill
Call For Price
COMPARE
0, 50, 100°C -20 Calibrated Liquid-In-Glass Thermometer; -20 to 110C, Total Immersion, Organic Liquid Fill
Call For Price
COMPARE
0, 60, 120°C -20 Calibrated Liquid-In-Glass Thermometer, -20 to 150°C, Total Immersion, Organic Liquid Fill
Call For Price
COMPARE
1, 100, 200°C -10 Calibrated Liquid-In-Glass Thermometer, -10 to 225°C, Total Immersion, Organic Liquid Fill
Call For Price
COMPARE
0, 50, 100°C -1 Calibrated Liquid-In-Glass Thermometer; -1 to 101C, Total Immersion, Organic Liquid Fill
Call For Price
COMPARE
0, 37, 56°C -20 Calibrated Liquid-In-Glass Thermometer, -20 to 110°C, 76 mm Immersion, Organic Liquid Fill, Calibration Points of 0, 37, and 56°C
₹56,000.00INR / Each
COMPARE
0, 10, 20, 30, 37, 40, 50°C -1 Calibrated Liquid-In-Glass Thermometer; -1 to 51C, 76mm Immersion, Organic Liquid Fill
₹73,400.00INR / Each
COMPARE
0, 10, 20, 30, 40, 50, 56, 60, 70, 80, 90, 100°C -1 Calibrated Liquid-In-Glass Thermometer; -1 to 101C, 76mm Immersion, Organic Liquid Fill
₹115,000.00INR / Each
COMPARE
0, 20, 37, 56, 80, 100, 121, 150°C -1 Calibrated Liquid-In-Glass Thermometer; -1 to 201C, 76mm Immersion, Organic Liquid Fill
₹59,100.00INR / Each
COMPARE
0, 10, 20, 30, 40, 50, 60, 70, 80, 90, 100°C -1 Calibrated Liquid-In-Glass Thermometer; -1 to 101C, Total Immersion, Organic Liquid Fill SKU 9030021
₹112,000.00INR / Each
COMPARE
0, 10, 20, 30, 37, 40, 50°C -1 Calibrated Liquid-In-Glass Thermometer; -1 to 51C, Total Immersion, Organic Liquid Fill
₹78,600.00INR / Each
COMPARE
0, 25, 37, 44.6, 100, 121, 140, 150, 160, 200°C -1 Calibrated Liquid-In-Glass Thermometer; -1 to 201C, Total Immersion, Organic Liquid Fill
₹112,000.00INR / Each
COMPARE
-40, -20, 0°C -50 Calibrated Liquid-In-Glass Thermometer; -50 to 50C, Total Immersion, Organic Liquid Fill
Call For Price
Showing 1- 17 of 17