Diba Omnifit® Inline Filters
Chemically inert flow path with all PTFE wetted parts
  • Simple filter replacement
Use PTFE inline filters with aggressive and high-purity fluids. Filters remove particulates from the medium protecting instruments downstream from particulate damage. Each filter housing holds a removable PTFE filter.
25,300.00 - ₹51,100.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connections Pore Size (micron)
Availability Pricing
Diba Omnifit® Inline Solvent Filter, PTFE, Cap System connection, 50 µm; 1/ea 2194019 Mfr # 003303
COMPARE
Cap-style connection with a flat-bottom 1/4"-28 port 50 µm
₹51,100.00INR / Each
Diba Omnifit® Inline Solvent Filter, PTFE, 1/4"-28 UNF(F) flat bottom, 20 µm; 1/ea 2194020 Mfr # 003305
COMPARE
1/4"-28 UNF(F) 20 µm
₹25,300.00INR / Each