Diba Omnifit® Inline Solvent Filter, PTFE, Cap System connection, 50 µm; 1/ea
Representative image only.
Diba Omnifit® Inline Solvent Filter, PTFE, Cap System connection, 50 µm; 1/ea
Diba | Mfr: 003303| Item: 2194019
Chemically inert flow path with all PTFE wetted parts
₹51,100.00INR / Each
₹51,100.00INR / Each
Buy It Together
Current Item
Diba Omnifit® Inline Solvent Filter, PTFE, Cap System connection, 50 µm; 1/ea
Diba Omnifit® Inline Solvent Filter, PTFE, Cap System connection, 50 µm; 1/ea
2194019
₹51,100.00INR / Each
+ =
Total Price 107300.00 ADD 2 ITEMS TO CART
Product Description
Includes
PTFE filter element
Key Features
  • Simple filter replacement
More About this Item
Use PTFE inline filters with aggressive and high-purity fluids. Filters remove particulates from the medium protecting instruments downstream from particulate damage. Each filter housing holds a removable PTFE filter.
Specifications
  • Membrane MaterialPTFE
  • Pore Size (micron)50 µm
  • Housing MaterialPTFE
  • ConnectionsCap-style connection with a flat-bottom 1/4"-28 port
  • Ports1/4"-28 port that accepts 1/16" to 1/8" rigid tubing