Diba Omnifit® Inline Solvent Filter, PTFE, 1/4"-28 UNF(F) flat bottom, 20 µm; 1/ea
Representative image only.
Diba Omnifit® Inline Solvent Filter, PTFE, 1/4"-28 UNF(F) flat bottom, 20 µm; 1/ea
Diba | Mfr: 003305| Item: 2194020
Chemically inert flow path with all PTFE wetted parts
₹25,300.00INR / Each
₹25,300.00INR / Each
Buy It Together
Current Item
Diba Omnifit® Inline Solvent Filter, PTFE, 1/4"-28 UNF(F) flat bottom, 20 µm; 1/ea
Diba Omnifit® Inline Solvent Filter, PTFE, 1/4"-28 UNF(F) flat bottom, 20 µm; 1/ea
2194020
₹25,300.00INR / Each
+ =
Total Price 50200.00 ADD 2 ITEMS TO CART
Product Description
Includes
PTFE filter element
Key Features
  • Simple filter replacement
More About this Item
Use PTFE inline filters with aggressive and high-purity fluids. Filters remove particulates from the medium protecting instruments downstream from particulate damage. Each filter housing holds a removable PTFE filter.
Specifications
  • Membrane MaterialPTFE
  • Pore Size (micron)20 µm
  • Housing MaterialPTFE
  • Connections1/4"-28 UNF(F)