Diba Omnifit® ETFE Adapters with Luer Connection
Convert Omnifit cap system fittings or 1/4"-28 UNF ports to Luer connectors
  • Constructed of durable ETFE
Cap adapter replaces the colored cap on the cap system fittings and valves to convert it to a male or female Luer connection. The 1/4"-28 UNF adapters fit on 1/4"-28 UNF flat bottom ports of cap system fittings and valves.
3,100.00 - ₹57,900.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connection Type-Port 1 Connection Type-Port 2
Availability Pricing
Diba Omnifit® Cap System Adapter, male Luer slip tip; 1/ea WW-21939-29 Mfr # 001201
COMPARE
Omnifit® cap system Male Luer slip
₹57,900.00INR / Each
Diba Omnifit® Cap System Adapter, female Luer; 1/ea WW-21939-30 Mfr # 001202
COMPARE
Omnifit® cap system Female Luer
₹3,800.00INR / Each
Diba Omnifit® Port Adapter, 1/4"-28 UNF(M) to Luer lock; 1/ea WW-21939-31 Mfr # 001206
COMPARE
1/4\"-28 UNF(M) Male Luer lock
₹8,000.00INR / Each
Diba Omnifit® Port Adapter, 1/4"-28 UNF(M) to female Luer; 1/ea WW-21940-11 Mfr # 002502
COMPARE
1/4\"-28 UNF(M) Female Luer
₹3,100.00INR / Each