Diba Omnifit® Port Adapter, 1/4"-28 UNF(M) to Luer lock; 1/ea
Representative image only.
Diba Omnifit® Port Adapter, 1/4"-28 UNF(M) to Luer lock; 1/ea
Diba | Mfr: 001206| Item: 2193931
Convert Omnifit cap system fittings or 1/4"-28 UNF ports to Luer connectors
₹7,600.00INR / Each
₹7,600.00INR / Each
Product Description
Key Features
  • Constructed of durable ETFE
More About this Item
Cap adapter replaces the colored cap on the cap system fittings and valves to convert it to a male or female Luer connection. The 1/4"-28 UNF adapters fit on 1/4"-28 UNF flat bottom ports of cap system fittings and valves.
Specifications
  • Length (")1 11/32
  • MaterialETFE
  • Width (InchShort)25/64
  • Number of Ports2
  • Connection Type-Port 11/4"-28 UNF(M)
  • Connection Type-Port 2Male Luer lock
  • DescriptionOmnifit® Port Adapter, 1/4"-28 UNF(M) to Luer lock; 1/ea