Diba Omnifit® Cap System Adapter, female Luer; 1/ea
Representative image only.
Diba Omnifit® Cap System Adapter, female Luer; 1/ea
Diba | Mfr: 001202| Item: 2193930
Convert Omnifit cap system fittings or 1/4"-28 UNF ports to Luer connectors
₹5,700.00INR / Each
₹5,700.00INR / Each
Product Description
Key Features
  • Constructed of durable ETFE
More About this Item
Cap adapter replaces the colored cap on the cap system fittings and valves to convert it to a male or female Luer connection. The 1/4"-28 UNF adapters fit on 1/4"-28 UNF flat bottom ports of cap system fittings and valves.
Specifications
  • Length (")1 9/64
  • MaterialETFE
  • Number of Ports2
  • Width (InchShort)25/64
  • Connection Type-Port 1Omnifit® cap system
  • Connection Type-Port 2Female Luer
  • DescriptionOmnifit® Cap System Adapter, female Luer; 1/ea