Diba Click-N-Seal® Fittings with Flat-Bottom 1/4"-28 UNF(M) Thread for Flared Tubing
Achieve a leak-free seal with only the tube material in the fluid path
  • When the cap clicks, you know the seal is tight
  • Self-torquing fittings create a secure seal without the use of any tools
Fittings for flared tubing create a ferrule-free connection and ensure leak-free seal while minimizing carryover. As the fitting is tightened, the resistance of the cap increases until the correct torque is reached. A seal is created by compressing the flared portion of the tube against the flat bottom of the port.
Select from acetal, polycarbonate (PC), or PEEK™ material of construction. PEEK fittings are compact in design and provide excellent chemical resistance. Choose the size that best matches your tube OD. A washer is recommended to ensure the flare seals correctly. Use flanging kit to easily flange semi-rigid tubing.
5,200.00 - ₹15,900.00INR / Pkg of 6
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Compression Size / Tube Od (Port 1) (in) Fitting Material
Availability Pricing
Diba Click-N-Seal™ Plus Tubing End Fitting, 1/16" OD flared tubing, 1/4"-28 UNF(M), white acetal; 6/pk WW-21942-40 Mfr # 008NG16-AT5H
COMPARE
White 1/16 Acetal
₹6,000.00INR / Pkg of 6
Diba Click-N-Seal™ Tubing End Fitting, 1/16" OD flared tubing, 1/4"-28 UNF(M), black PC; 6/pk WW-21942-42 Mfr # 008NG16-CS5B
COMPARE
Black 1/16 Polycarbonate
₹5,200.00INR / Pkg of 6
Diba Click-N-Seal™ Tubing End Fitting, 1/16" OD flared tubing, 1/4"-28 UNF(M), white PC; 10/pk WW-21942-43 Mfr # 008NG16-CS5H
COMPARE
White 1/16 Polycarbonate
₹5,200.00INR / Pkg of 10
Diba Click-N-Seal™ Tubing End Fitting, 1/16" OD flared tubing, 1/4"-28 UNF(M), red PC; 6/pk WW-21942-44 Mfr # 008NG16-CS5R
COMPARE
Red 1/16 Polycarbonate
₹5,200.00INR / Pkg of 6
Diba Click-N-Seal™ Ultra Tubing End Fitting, 1/16" OD flared tubing, 1/4"-28 UNF(M), natural PEEK™; 6/pk WW-21942-48 Mfr # 008NG16-KT5L
COMPARE
Natural 1/16 PEEK
₹15,900.00INR / Pkg of 6
Diba Click-N-Seal™ Plus Tubing End Fitting, 1.6 to 2.5mm OD flared tubing, 1/4"-28 UNF(M), acetal; 6/pk WW-21942-49 Mfr # 008NG25-AT5H
COMPARE
White 3/32 Acetal
₹7,900.00INR / Pkg of 6
Diba Click-N-Seal™ Tubing End Fitting, 1.6 to 2.5mm OD flared tubing, 1/4"-28 UNFM, black PC; 6/pk WW-21942-51 Mfr # 008NG25-CS5B
COMPARE
Black 3/32 Polycarbonate
₹5,200.00INR / Pkg of 6
Diba Click-N-Seal™ Tubing End Fitting, 1.6 to 2.5mm OD flared tubing, 1/4"-28 UNFM, white PC; 6/pk WW-21942-52 Mfr # 008NG25-CS5H
COMPARE
White 3/32 Polycarbonate
₹5,200.00INR / Pkg of 6
Diba Click-N-Seal™ Tubing End Fitting, 1.6 to 2.5mm OD flared tubing, 1/4"-28 UNFM, red PC; 6/pk WW-21942-53 Mfr # 008NG25-CS5R
COMPARE
Red 3/32 Polycarbonate
₹5,200.00INR / Pkg of 6
Diba Click-N-Seal™ Ultra Tubing End Fitting, 1.6 to 2.5mm OD flared tubing, 1/4"-28 UNFM, PEEK; 6/pk WW-21942-57 Mfr # 008NG25-KT5L
COMPARE
Natural 3/32 PEEK
₹15,900.00INR / Pkg of 6
Diba Click-N-Seal™ Tubing End Fitting, 1/8" OD flared tubing, 1/4"-28 UNF(M), white acetal; 6/pk WW-21942-58 Mfr # 008NG32-AT5H
COMPARE
White 1/8 Acetal
₹7,900.00INR / Pkg of 6
Diba Click-N-Seal™ Tubing End Fitting, 1/8" OD flared tubing, 1/4"-28 UNF(M), black PC; 6/pk WW-21942-60 Mfr # 008NG32-CS5B
COMPARE
Black 1/8 Polycarbonate
₹5,200.00INR / Pkg of 6
Diba Click-N-Seal™ Tubing End Fitting, 1/8" OD flared tubing, 1/4"-28 UNF(M), white PC; 6/pk WW-21942-61 Mfr # 008NG32-CS5H
COMPARE
White 1/8 Polycarbonate
₹5,200.00INR / Pkg of 6
Diba Click-N-Seal™ Tubing End Fitting, 1/8" OD flared tubing, 1/4"-28 UNF(M), red PC; 6/pk WW-21942-62 Mfr # 008NG32-CS5R
COMPARE
Red 1/8 Polycarbonate
₹5,200.00INR / Pkg of 6
Diba Click-N-Seal™ Tubing End Fitting, 1/8" OD flared tubing, 1/4"-28 UNF(M), natural PEEK™; 6/pk WW-21942-66 Mfr # 008NG32-KT5L
COMPARE
Natural 1/8 PEEK
₹15,200.00INR / Pkg of 6
Showing 1- 15 of 15