Diba Click-N-Seal™ Tubing End Fitting, 1.6 to 2.5mm OD flared tubing, 1/4"-28 UNFM, black PC; 6/pk
Representative image only.
Diba Click-N-Seal™ Tubing End Fitting, 1.6 to 2.5mm OD flared tubing, 1/4"-28 UNFM, black PC; 6/pk
Diba | Mfr: 008NG25-CS5B| Item: 2194251
Achieve a leak-free seal with only the tube material in the fluid path
₹10,000.00INR / Pkg of 6
₹10,000.00INR / Pkg of 6
Buy It Together
Current Item
Diba Click-N-Seal™ Tubing End Fitting, 1.6 to 2.5mm OD flared tubing, 1/4"-28 UNFM, black PC; 6/pk
Diba Click-N-Seal™ Tubing End Fitting, 1.6 to 2.5mm OD flared tubing, 1/4"-28 UNFM, black PC; 6/pk
2194251
₹10,000.00INR / Pkg of 6
+ =
Total Price 169000.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • When the cap clicks, you know the seal is tight
  • Self-torquing fittings create a secure seal without the use of any tools
More About this Item
Fittings for flared tubing create a ferrule-free connection and ensure leak-free seal while minimizing carryover. As the fitting is tightened, the resistance of the cap increases until the correct torque is reached. A seal is created by compressing the flared portion of the tube against the flat bottom of the port.
Select from acetal, polycarbonate (PC), or PEEK™ material of construction. PEEK fittings are compact in design and provide excellent chemical resistance. Choose the size that best matches your tube OD. A washer is recommended to ensure the flare seals correctly. Use flanging kit to easily flange semi-rigid tubing.
An O-ring may be necessary to ensure the fitting seats correctly in deeper ports.
Specifications
  • Fitting MaterialPolycarbonate
  • Connection TypeCompression x UNF(M)
  • Thread size UNF(M) (port 2) (InchShort)1/4-28
  • Compression Size / Tube Od (Port 1) (in)3/32
  • Compression Size / Tube Od (Port 1) (mm)2.5
  • ColorBlack
  • DescriptionClick-N-Seal™ Tubing End Fitting, 1.6 to 2.5mm OD flared tubing, 1/4"-28 UNFM, black PC; 6/pk