Diba Omnifit® Flanging Kit for Rigid Tubing, 1/16" or 1/8" OD tubing
Representative image only.
Diba Omnifit® Flanging Kit for Rigid Tubing, 1/16" or 1/8" OD tubing
Diba | Mfr: 007003| Item: 2194023
No electrical power required!
₹159,000.00INR / Each
₹159,000.00INR / Each
Product Description
Key Features
  • Suitable for use with Click-N-Seal® fittings for flared tubing
  • Flange semi-rigid tubing without electrical power or heat
  • Works on tubing with 1/16" to 1/8" tube OD
More About this Item
Use the flanging tool to create a flange on rigid tubing with 1/16" to 1/8" OD.
Specifications
  • Compatible WithClick-N-Seal® Fittings for Flared Tubing
  • DescriptionOmnifit® Flanging Kit for Rigid Tubing, 1/16" or 1/8" OD tubing