Diba Click-N-Seal™ Tubing End Fitting, 1/16" OD flared tubing, 1/4"-28 UNF(M), white PC; 10/pk
Representative image only.
Diba Click-N-Seal™ Tubing End Fitting, 1/16" OD flared tubing, 1/4"-28 UNF(M), white PC; 10/pk
Diba | Mfr: 008NG16-CS5H| Item: 2194243
Achieve a leak-free seal with only the tube material in the fluid path
₹10,000.00INR / Pkg of 10
₹10,000.00INR / Pkg of 10
Buy It Together
Current Item
Diba Click-N-Seal™ Tubing End Fitting, 1/16" OD flared tubing, 1/4"-28 UNF(M), white PC; 10/pk
Diba Click-N-Seal™ Tubing End Fitting, 1/16" OD flared tubing, 1/4"-28 UNF(M), white PC; 10/pk
2194243
₹10,000.00INR / Pkg of 10
+ =
Total Price 169000.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • When the cap clicks, you know the seal is tight
  • Self-torquing fittings create a secure seal without the use of any tools
More About this Item
Fittings for flared tubing create a ferrule-free connection and ensure leak-free seal while minimizing carryover. As the fitting is tightened, the resistance of the cap increases until the correct torque is reached. A seal is created by compressing the flared portion of the tube against the flat bottom of the port.
Select from acetal, polycarbonate (PC), or PEEK™ material of construction. PEEK fittings are compact in design and provide excellent chemical resistance. Choose the size that best matches your tube OD. A washer is recommended to ensure the flare seals correctly. Use flanging kit to easily flange semi-rigid tubing.
An O-ring may be necessary to ensure the fitting seats correctly in deeper ports.
Specifications
  • Fitting MaterialPolycarbonate
  • Connection TypeCompression x UNF(M)
  • Thread size UNF(M) (port 2) (InchShort)1/4-28
  • Compression Size / Tube Od (Port 1) (in)1/16
  • Compression Size / Tube Od (Port 1) (mm)1.57
  • ColorWhite
  • DescriptionClick-N-Seal™ Tubing End Fitting, 1/16" OD flared tubing, 1/4"-28 UNF(M), white PC; 10/pk