Diba Click-N-Seal® Fitings with 10-32 UNF(M) Fittings for 1/16" OD Tubing
Quick and easy connection of 1/16" OD tubing into a 10-32 port
  • Hand tighten fittings without tools to create a secure seal
  • When the cap clicks, you know the seal is tight
Fittings are constructed of PEEK material. Select from conical bottom or flat bottom 10-32 UNF(M) fittings. Conical bottom fittings withstand up to 2000 psi and are suitable for HPLC type ports. Order type S conical bottom ferrules 21940-53 to use with flat bottom fittings.
17,700.00INR / Pkg of 10
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Thread size UNF(M) (port 2) (InchShort) Description
Availability Pricing
Diba Click-N-Seal™ Tubing End Fitting, Type S/P, 1/16" OD tubing, 10-32 UNF(M), Flat Bottom, PEEK; 10/pk 2194201 Mfr # 008NF16-KT4L
COMPARE
10-32 Click-N-Seal™ Tubing End Fitting, Type S/P, 1/16" OD tubing, 10-32 UNF(M), Flat Bottom, PEEK; 10/pk
₹17,700.00INR / Pkg of 10
Diba Click-N-Seal™ Tubing End Fitting, 1/16" OD flared tubing, 10-32 UNF(M), Conical Bottom, natural PEEK™; 6/pk 2194267 Mfr # 008NT16-KT4L
COMPARE
10-32 Click-N-Seal™ Tubing End Fitting, 1/8" OD flared tubing, 10-32 UNF(M), Conical Bottom, natural PEEK™; 6/pk
₹17,700.00INR / Pkg of 6