Diba Click-N-Seal™ Tubing End Fitting, Type S/P, 1/16" OD tubing, 10-32 UNF(M), Flat Bottom, PEEK; 10/pk
Representative image only.
Diba Click-N-Seal™ Tubing End Fitting, Type S/P, 1/16" OD tubing, 10-32 UNF(M), Flat Bottom, PEEK; 10/pk
Diba | Mfr: 008NF16-KT4L| Item: 2194201
Quick and easy connection of 1/16" OD tubing into a 10-32 port
₹17,700.00INR / Pkg of 10
₹17,700.00INR / Pkg of 10
Buy It Together
Current Item
Diba Click-N-Seal™ Tubing End Fitting, Type S/P, 1/16" OD tubing, 10-32 UNF(M), Flat Bottom, PEEK; 10/pk
Diba Click-N-Seal™ Tubing End Fitting, Type S/P, 1/16" OD tubing, 10-32 UNF(M), Flat Bottom, PEEK; 10/pk
2194201
₹17,700.00INR / Pkg of 10
+ =
Total Price 31000.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Hand tighten fittings without tools to create a secure seal
  • When the cap clicks, you know the seal is tight
More About this Item
Fittings are constructed of PEEK material. Select from conical bottom or flat bottom 10-32 UNF(M) fittings. Conical bottom fittings withstand up to 2000 psi and are suitable for HPLC type ports. Order type S conical bottom ferrules 21940-53 to use with flat bottom fittings.
Specifications
  • Fitting MaterialPEEK
  • Connection TypeCompression x UNF(M)
  • Thread size UNF(M) (port 2) (InchShort)10-32
  • Compression Size / Tube Od (Port 1) (in)1/16
  • Compression Size / Tube Od (Port 1) (mm)1.57
  • ColorNatural
  • Max Operating Pressure (PSI)100
  • DescriptionClick-N-Seal™ Tubing End Fitting, Type S/P, 1/16" OD tubing, 10-32 UNF(M), Flat Bottom, PEEK; 10/pk