Diba Click-N-Seal™ Tubing End Fitting, 1/16" OD flared tubing, 10-32 UNF(M), Conical Bottom, natural PEEK™; 6/pk
Representative image only.
Diba Click-N-Seal™ Tubing End Fitting, 1/16" OD flared tubing, 10-32 UNF(M), Conical Bottom, natural PEEK™; 6/pk
Diba | Mfr: 008NT16-KT4L| Item: 2194267
Quick and easy connection of 1/16" OD tubing into a 10-32 port
₹17,700.00INR / Pkg of 6
₹17,700.00INR / Pkg of 6
Product Description
Key Features
  • Hand tighten fittings without tools to create a secure seal
  • When the cap clicks, you know the seal is tight
More About this Item
Fittings are constructed of PEEK material. Select from conical bottom or flat bottom 10-32 UNF(M) fittings. Conical bottom fittings withstand up to 2000 psi and are suitable for HPLC type ports. Order type S conical bottom ferrules 21940-53 to use with flat bottom fittings.
Specifications
  • Fitting MaterialPEEK
  • Connection TypeCompression x UNF(M)
  • Thread size UNF(M) (port 2) (InchShort)10-32
  • Compression Size / Tube Od (Port 1) (in)1/16
  • Compression Size / Tube Od (Port 1) (mm)1.57
  • ColorNatural
  • Max Operating Pressure (PSI)2000
  • DescriptionClick-N-Seal™ Tubing End Fitting, 1/8" OD flared tubing, 10-32 UNF(M), Conical Bottom, natural PEEK™; 6/pk