Culture Tube Closures
  • Closures minimize evaporation while allowing necessary gas exchange.
  • Positive-grip closures of stainless steel or autoclavable polypropylene.
These new tube closures ensure the viability of microbiological cultures by protecting solid and broth media from excessive evaporation. They fit snugly on rimless culture tubes and flasks while allowing for gas exchange and ventilation.
70,800.00 - ₹168,000.00INR / Case of 144
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Kap-Uts 2005-00013 Culture tube closure, 13 mm, stainless steel 0450000 Mfr # 2005-00013A
COMPARE
Culture tube closure, 13 mm, stainless steel
₹150,000.00INR / Case of 144
Kap-Uts 2005-00016 Culture tube closure, 16 mm, stainless steel 0450001 Mfr # 2005-00016
COMPARE
Culture tube closure, 16 mm, stainless steel
₹160,000.00INR / Case of 144
Kap-Uts 2005-00018 Culture tube closure, 18 mm, stainless steel 0450002 Mfr # 2005-00018
COMPARE
Culture tube closure, 18 mm, stainless steel
₹168,000.00INR / Case of 144
Kap-Uts 2005-00025 Culture tube closure, 25 mm, stainless steel 0450003 Mfr # 2005-00025
COMPARE
Culture tube closure, 25 mm, stainless steel
₹116,000.00INR / Case of 72
Kap-Uts 2007-13005 Polypropylene Culture Tube Closures, 13 mm, white 0450100 Mfr # 2007-13005
COMPARE
Polypropylene Culture Tube Closures, 13 mm, white
₹100,000.00INR / Case of 1000
Kap-Uts 2007-16001 Polypropylene Culture Tube Closures, 16 mm, black 0450101 Mfr # 2007-16001
COMPARE
Polypropylene Culture Tube Closures, 16 mm, black
₹141,000.00INR / Case of 1000
Kap-Uts 2007-16002 Polypropylene Culture Tube Closures, 16 mm, red 0450102 Mfr # 2007-16002
COMPARE
Polypropylene Culture Tube Closures, 16 mm, red
₹141,000.00INR / Case of 1000
Kap-Uts 2007-16003 Polypropylene Culture Tube Closures, 16 mm, yellow 0450103 Mfr # 2007-16003
COMPARE
Polypropylene Culture Tube Closures, 16 mm, yellow
₹168,000.00INR / Case of 1000
Kap-Uts 2007-16004 Polypropylene Culture Tube Closures, 16 mm, blue 0450104 Mfr # 2007-16004
COMPARE
Polypropylene Culture Tube Closures, 16 mm, blue
₹141,000.00INR / Case of 1000
Kap-Uts 2007-16005 Polypropylene Culture Tube Closures, 16 mm, white 0450105 Mfr # 2007-16005
COMPARE
Polypropylene Culture Tube Closures, 16 mm, white
₹138,000.00INR / Case of 1000
Kap-Uts 2007-25005 Polypropylene Culture Tube Closures, 25 mm, white 0450108 Mfr # 2007-25005
COMPARE
Polypropylene Culture Tube Closures, 25 mm, white
₹70,800.00INR / Case of 500
Showing 1- 11 of 11