Kap-Uts 2007-16004 Polypropylene Culture Tube Closures, 16 mm, blue
Representative image only.
Kap-Uts 2007-16004 Polypropylene Culture Tube Closures, 16 mm, blue
Kap-Uts | Mfr: 2007-16004| Item: 0450104
₹141,000.00INR / Case of 1000
₹141,000.00INR / Case of 1000
Product Description
Key Features
  • Closures minimize evaporation while allowing necessary gas exchange.
  • Positive-grip closures of stainless steel or autoclavable polypropylene.
More About this Item
These new tube closures ensure the viability of microbiological cultures by protecting solid and broth media from excessive evaporation. They fit snugly on rimless culture tubes and flasks while allowing for gas exchange and ventilation.
Specifications
  • DescriptionPolypropylene Culture Tube Closures, 16 mm, blue