Kap-Uts 2005-00018 Culture tube closure, 18 mm, stainless steel
Representative image only.
Kap-Uts 2005-00018 Culture tube closure, 18 mm, stainless steel
Kap-Uts | Mfr: 2005-00018| Item: 0450002
₹168,000.00INR / Case of 144
₹168,000.00INR / Case of 144
Product Description
Key Features
  • Closures minimize evaporation while allowing necessary gas exchange.
  • Positive-grip closures of stainless steel or autoclavable polypropylene.
More About this Item
These new tube closures ensure the viability of microbiological cultures by protecting solid and broth media from excessive evaporation. They fit snugly on rimless culture tubes and flasks while allowing for gas exchange and ventilation.
Specifications
  • DescriptionCulture tube closure, 18 mm, stainless steel