CPC® (Colder) EFC Quick-Disconnect Fittings, Hose Barb Body & Insert, Polypropylene
High efficiency valve provides more flow in a compact size
  • Ideal for general purposes, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support is available to determine which connector type is needed for your application
  • Chemically resistant and gamma sterilizable
  • Ideal for tubing sizes from 1/4“ to 3/8” ID
  • Compatible with most APC couplings
The CPC (Colder) EFC Series quick-disconnect fittings offer a high-efficiency valve design, delivering superior flow capacity in a compact size. Featuring a plastic thumb latch with no exposed metal, these fittings ensure easy, gloved-hand operation. Made from chemically resistant polypropylene, they are ideal for harsh environments, providing durability and reliable performance.
720.00 - ₹2,800.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Flow Pattern Connection Type
Availability Pricing
CPC (Colder) EFC12 Quick-Disconnect  Fittings, Hose Barb Insert, Polypropylene, Non-Valved, 1/4" ID; 1/Each 5013519 Mfr # EFC22412
COMPARE
1/4 Straight Quick-Disconnect Hose Barb Insert, Non-Valve
₹720.00INR / Each
CPC (Colder) EFC12 Quick-Disconnect  Fittings, Hose Barb Body, Polypropylene, Valved, 1/4" ID; 1/Each 5013520 Mfr # EFCD17412
COMPARE
1/4 Straight Quick-Disconnect Hose Barb Body, Valve
₹2,800.00INR / Each
CPC (Colder) EFC12 Quick-Disconnect  Fittings, Hose Barb Insert, Polypropylene, Valved, 1/4" ID; 1/Each 5013521 Mfr # EFCD22412
COMPARE
1/4 Straight Quick-Disconnect Hose Barb Insert, Valve
₹2,400.00INR / Each