Control Products Temperature Switches
Use to control system temperatures or to act as a final safety shutoff
Designed for use in rigorous industrial or laboratory condition to control overheating of air compressors, distilling machines, and other equipment. A bimetal snap disc reverses from convex to concave upon reaching temperature trip point, which activates a SPDT microswitch.
The three-wire design allows you to wire SPDT switch for either normally open (NO) or normally closed (NC) operation. Simple to install—switches require very little wiring and don't need shock mounts. Compact enough to easily thread into any 1/2" NPT opening. Stainless steel housing.
81,800.00 - ₹82,400.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Diameter (in) Length (in) Max Temperature (° F)
Availability Pricing
Control Products AD148-12/100F Temperature Switches Temperature Trip Point; 100F 9388052 Mfr # AD148-12/100F
COMPARE
11/16 1 1/4 100
₹81,800.00INR / Each
Control Products AD148-12/125F Temperature SwitchesTemperature Trip Point; 125F 9388054 Mfr # AD148-12/125F
COMPARE
11/16 1 1/4 125
₹82,400.00INR / Each
Control Products AD148-12/150F Temperature SwitchesTemperature Trip Point; 150F 9388056 Mfr # AD148-12/150F
COMPARE
11/16 1 1/4 150
₹82,400.00INR / Each
Control Products AD148-12/250F Temperature Switches; temperature trip point; 250F 9388062 Mfr # AD148-12/250F
COMPARE
11/16 1 1/4 250
₹81,800.00INR / Each