Control Products AD148-12/250F Temperature Switches; temperature trip point; 250F
Representative image only.
Control Products AD148-12/250F Temperature Switches; temperature trip point; 250F
Control Products | Mfr: AD148-12/250F| Item: 9388062
Use to control system temperatures or to act as a final safety shutoff
₹81,800.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹81,800.00INR / Each
Buy It Together
Current Item
Control Products AD148-12/250F Temperature Switches; temperature trip point; 250F
Control Products AD148-12/250F Temperature Switches; temperature trip point; 250F
9388062
₹81,800.00INR / Each
+ =
Total Price 126500.00 ADD 2 ITEMS TO CART
Product Description
More About this Item
Designed for use in rigorous industrial or laboratory condition to control overheating of air compressors, distilling machines, and other equipment. A bimetal snap disc reverses from convex to concave upon reaching temperature trip point, which activates a SPDT microswitch.
The three-wire design allows you to wire SPDT switch for either normally open (NO) or normally closed (NC) operation. Simple to install—switches require very little wiring and don't need shock mounts. Compact enough to easily thread into any 1/2" NPT opening. Stainless steel housing.
Specifications
  • Min Temperature (° F)250
  • Max Temperature (° F)250
  • Length (in)1 1/4
  • Diameter (in)11/16
  • DescriptionTemperature Switches; temperature trip point; 250F