Colitag™ Water Test Kits
Quickly and confidently analyze drinking water
  • Colitag is a presence-absence and MPN enzyme substrate test
  • Detects as few as 1 MPN of total coliform or E. coli in a 100-mL water sample
  • US EPA approved; depended on in over 24 countries
  • Maximizes flexibility; use one test instead of three
Enhance your laboratory efficiency—and ensure water purity—with Colitag, the revolutionary 16-48 hour water test. To use, simply add Colitag to a 100-mL sample, incubate, visually check for coliforms and illuminate with a UV lamp, as bright blue fluorescence indicates a positive E. coli sample.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Test Tests Per Kit Description
Availability Pricing
Neogen COLI9851 Colitag™ Test Kit, P/A 100-mL Format; 100/Pk 5320017 Mfr # COLI9851
COMPARE
Call For Price
Neogen COLI9857 Colitag™ Comparator, P/A 100-mL Format; 1/Ea 5320025 Mfr # COLI9857
COMPARE
Coliform, E. coli 1 Colitag™ Comparator, P/A 100-mL Format; 1/Ea