Neogen COLI9851 Colitag™ Test Kit, P/A 100-mL Format; 100/Pk
Representative image only.
Neogen COLI9851 Colitag™ Test Kit, P/A 100-mL Format; 100/Pk
| Mfr: COLI9851| Item: 5320017
Quickly and confidently analyze drinking water
Call For Price
Call For Price
Buy It Together
Current Item
Neogen COLI9851 Colitag™ Test Kit, P/A 100-mL Format; 100/Pk
Neogen COLI9851 Colitag™ Test Kit, P/A 100-mL Format; 100/Pk
5320017
Call For Price
+ =
Total Price 0 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Colitag is a presence-absence and MPN enzyme substrate test
  • Detects as few as 1 MPN of total coliform or E. coli in a 100-mL water sample
  • US EPA approved; depended on in over 24 countries
  • Maximizes flexibility; use one test instead of three
More About this Item
Enhance your laboratory efficiency—and ensure water purity—with Colitag, the revolutionary 16-48 hour water test. To use, simply add Colitag to a 100-mL sample, incubate, visually check for coliforms and illuminate with a UV lamp, as bright blue fluorescence indicates a positive E. coli sample.