Neogen COLI9857 Colitag™ Comparator, P/A 100-mL Format; 1/Ea
Representative image only.
Neogen COLI9857 Colitag™ Comparator, P/A 100-mL Format; 1/Ea
| Mfr: COLI9857| Item: 5320025
Quickly and confidently analyze drinking water
Call For Price
Call For Price
Buy It Together
Current Item
Neogen COLI9857 Colitag™ Comparator, P/A 100-mL Format; 1/Ea
Neogen COLI9857 Colitag™ Comparator, P/A 100-mL Format; 1/Ea
5320025
Call For Price
+ =
Total Price 0 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Colitag is a presence-absence and MPN enzyme substrate test
  • Detects as few as 1 MPN of total coliform or E. coli in a 100-mL water sample
  • US EPA approved; depended on in over 24 countries
  • Maximizes flexibility; use one test instead of three
More About this Item
Enhance your laboratory efficiency—and ensure water purity—with Colitag, the revolutionary 16-48 hour water test. To use, simply add Colitag to a 100-mL sample, incubate, visually check for coliforms and illuminate with a UV lamp, as bright blue fluorescence indicates a positive E. coli sample.
Specifications
  • TestColiform, E. coli
  • Tests Per Kit1
  • DescriptionColitag™ Comparator, P/A 100-mL Format; 1/Ea