Cole-Parmer® Wide Mouth Bottles, PTFE
Can be used with most chemical compounds and corrosive environments
These leakproof PTFE bottles are ideal for low-temperature applications up to 540°F (280°C) and should not be used with pressure applications. Bottles are isostatically molded to provide a smooth internal finish. Chemically resistant and inert; can be used with most chemical compounds and corrosive environments. Autoclavable.
5,800.00 - ₹80,400.00INR / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap / Stopper Size Material
Availability Pricing
COMPARE
480 90 mm Polytetrafluoroethylene (PTFE)
₹61,600.00INR / Each
COMPARE
1 10 mm Polytetrafluoroethylene (PTFE)
₹5,800.00INR / Each
COMPARE
5 10 mm Polytetrafluoroethylene (PTFE)
₹8,400.00INR / Each
COMPARE
10 12 mm Polytetrafluoroethylene (PTFE)
₹11,900.00INR / Each
COMPARE
25 19 mm Polytetrafluoroethylene (PTFE)
₹15,000.00INR / Each
COMPARE
50 25 mm Polytetrafluoroethylene (PTFE)
₹15,100.00INR / Each
COMPARE
100 35 mm Polytetrafluoroethylene (PTFE)
₹26,700.00INR / Each
COMPARE
150 35 mm Polytetrafluoroethylene (PTFE)
₹28,700.00INR / Each
COMPARE
250 42 mm Polytetrafluoroethylene (PTFE)
₹34,400.00INR / Each
COMPARE
500 52 mm Polytetrafluoroethylene (PTFE)
₹54,900.00INR / Each
COMPARE
1000 57 mm Polytetrafluoroethylene (PTFE)
₹80,400.00INR / Each
Showing 1- 11 of 11