Cole-Parmer® Wash Bottles, Integrated Spout
Fill up faster with wide-mouth design
  • No need to shake or tip to dispense sample
  • Increase flow by trimming tip
  • One-piece leakproof design with polypropylene screw closure
Dispense without tipping or shaking. Dispensing tip delivers uniform ultrafine stream and can be trimmed to increase flow. Bottle features a one-piece leakproof dispensing tube and PP screw closure. Meets US FDA 21 CFR regulation..
4,900.00 - ₹7,400.00INR / Pkg of 6
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap / Stopper Size Material
Availability Pricing
COMPARE
125 24 mm Low Density Polyethyene (LDPE)
₹7,400.00INR / Pkg of 6
COMPARE
250 38 mm Low Density Polyethyene (LDPE)
₹4,900.00INR / Pkg of 4
COMPARE
500 38 mm Low Density Polyethyene (LDPE)
₹5,800.00INR / Pkg of 4
COMPARE
750 38 mm Low Density Polyethyene (LDPE)
₹6,400.00INR / Pkg of 4