Cole-Parmer® Testing Sieves, 12" dia, Brass Frame/Brass Wire
These brass testing sieves are serialized and come with a test sieve certificate of compliance. Equivalent to ASTM E11-09 compliance sieve.
31,100.00 - ₹70,400.00INR / Each
26 variations of this product are available.
See All Product Options Below
26 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (InchShort) Sieve Opening (mm) U.S. Standard Screen Size (No.)
Availability Pricing
COMPARE
3.25 2.36 8
₹33,500.00INR / Each
COMPARE
3.25 2 10
₹33,500.00INR / Each
COMPARE
3.25 1.68 12
₹33,500.00INR / Each
COMPARE
3.25 1.41 14
₹33,500.00INR / Each
COMPARE
3.25 1.19 16
₹33,500.00INR / Each
COMPARE
3.25 1 18
₹33,500.00INR / Each
COMPARE
3.25 0.841 20
₹33,500.00INR / Each
COMPARE
3.25 0.707 25
₹33,500.00INR / Each
COMPARE
3.25 0.595 30
₹33,500.00INR / Each
COMPARE
3.25 0.5 35
₹33,500.00INR / Each
COMPARE
3.25 0.425 40
₹33,500.00INR / Each
COMPARE
3.25 0.297 50
₹33,500.00INR / Each
COMPARE
3.25 0.25 60
₹33,500.00INR / Each
COMPARE
3.25 0.212 70
₹33,500.00INR / Each
COMPARE
3.25 0.149 100
₹33,500.00INR / Each
COMPARE
3.25 0.105 140
₹39,400.00INR / Each
COMPARE
3.25 0.088 170
₹42,900.00INR / Each
COMPARE
3.25 0.074 200
₹46,900.00INR / Each
COMPARE
3.25 0.044 325
₹70,400.00INR / Each
COMPARE
1.625 2 10
₹31,100.00INR / Each
Showing 1 - 20 of 26