InnoCal NIST-Traceable Calibration; Sieve
Representative image only.
InnoCal NIST-Traceable Calibration; Sieve
InnoCal | Item: 1705050
NIST-traceable calibrations for sieves.
₹37,400.00INR / Each
₹37,400.00INR / Each
Product Description
Key Features
  • NIST-Traceable calibration for sieves
More About this Item
Calibration reports issued by InnoCal may not be reproduced, except in full, without written permission of InnoCal. The results stated in our reports relate only to the items tested or calibrated. Measurements reported on our reports are traceable to SI units via national standards maintained by NIST and are performed in compliance with MIL-STD-45662A, ANSI/NCSL Z540-1-1994, 10CFR50, Appendix B, ISO9002-94, and ISO/IEC 17025:2017.
By utilizing a sophisticated image analyzer traceable to the National Institute of Standards and Technology (NIST), your sieve is scanned for irregularities. Physical measurements are taken in a midpoint orientation of the wire in the X and y directions (warp and weft), and the wire diameters are measured to ensure compliance with the current ASTM E11 specification. The data gathered is summarized in a comprehensive verification report.
Specifications
  • Calibration TypeMechanical
  • System Calibration (Instrument and Probe)Yes
  • DescriptionNIST-Traceable Calibration Service; Sieve