Cole-Parmer® Syringe Filters
Minimize sample loss—designed for low hold-up
  • Filter culture media to organic solvents—variety membrane materials cover a wide range of applications
  • Depend on quality—all filters are tested to withstand pressures up to 75 psi (5.1 bar)
  • Sterile versions are certified RNase/DNase free
Choose the membrane material best suited for your application. Each filter is quality tested for filter efficiency and housing integrity for use up to 75 psi (5.1 bar). Sterile filters are certified nonpyrogenic, have been gamma sterilized, and come individually packaged with lot numbers printed on for easy traceability.
7,400.00 - ₹21,000.00INR / Pkg of 50
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Membrane Diameter (mm) Membrane Material Pore Size (µm)
Availability Pricing
COMPARE
30 Surfactant-Free Cellulose Acetate (SFCA) 0.22
Call For Price
COMPARE
30 Surfactant-Free Cellulose Acetate (SFCA) 0.45
₹21,000.00INR / Pkg of 50
COMPARE
30 Mixed cellulose ester (MCE) 0.22
₹7,800.00INR / Pkg of 45
COMPARE
30 Mixed cellulose ester (MCE) 0.45
₹7,800.00INR / Pkg of 45
COMPARE
13 Nylon 0.45
₹7,400.00INR / Pkg of 100
COMPARE
30 Nylon 0.45
₹7,600.00INR / Pkg of 50
COMPARE
30 Nylon 0.22
₹7,800.00INR / Pkg of 45
COMPARE
13 Polyethersulfone (PES) 0.22
₹8,700.00INR / Pkg of 100
COMPARE
13 Polyethersulfone (PES) 0.45
₹8,700.00INR / Pkg of 100
COMPARE
30 Polyethersulfone (PES) 0.22
₹7,800.00INR / Pkg of 45
COMPARE
30 Polyethersulfone (PES) 0.45
₹7,800.00INR / Pkg of 45
COMPARE
30 PTFE 0.45
₹7,600.00INR / Pkg of 45
COMPARE
30 PTFE 0.45
₹7,600.00INR / Pkg of 50
COMPARE
13 PVDF 0.22
₹8,500.00INR / Pkg of 100
COMPARE
30 PVDF 0.22
₹7,800.00INR / Pkg of 45
COMPARE
30 PVDF 0.45
₹7,800.00INR / Pkg of 45
Showing 1- 16 of 16