Cole-Parmer® Suspension Pack Insulated Shippers
Reusable and Recyclable container compatible with 2D bags from all manufacturers
  • Reusable and Recyclable container compatible with 2D bags from all manufacturers
  • A unique suspension pack to protect 2-D bags in a universal storage container during handling,freezing, storage and transportation
  • Equipped with UV protection shell and film to protect sensitive products
  • Stackable design increases efficiency and reduces wasted space
  • Available for varying sized bags that can be loaded into the same shell at any fill level
  • Versatile container with room for data loggers, other accessories (15960-78, 15960-79)
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Suspension Pack, 8 L
Call For Price
COMPARE
Suspension Pack, 20 L
Call For Price
COMPARE
Suspension Pack Insulated Shipper, 8 L
Call For Price
COMPARE
Suspension Pack Insulated Shipper, 20 L