Cole-Parmer® Sticky Roller and Handle
Easily remove dirt particles in your Cleanroom environment
Features:
• Knitted microfiber fabric and woven microfiber fabric
• Good sorbent of water and oil
• Ideal for automatically cleaning equipment and tool surface of LCD/FPD/PCB/ photoelectron/ semiconductor industry etc.
• Packaging: Double bagged, vacuum packaging
12,800.00 - ₹108,000.00INR / Case of 20
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (in) Material Description
Availability Pricing
COMPARE
3 LDPE Sticky Roller, 7.36", 6" to 10" W; 20bags/Cs
₹108,000.00INR / Case of 20
COMPARE
Non-contaminating SS and Plastic Handle for 6" Sticky Roller; 50sets/Cs
₹12,800.00INR / Case of 50