Cole-Parmer® Stainless Steel Fermentation Dissolved Oxygen Probes
Designed for fermentation applications where probes are repeatedly steam-sterilized
  • 316 SS Construction
  • Polarographic
All models feature a polarographic sensor, and are available in a variety of body diameters, mounting styles, and immersion lengths. Temperature compensated probes feature 100 Ω RTD temperature measuring sensors. Order cables separately.
175,000.00 - ₹210,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Fermentation DO Probe; SS/Polarographic/320 mm, 3/4" NPT
₹210,000.00INR / Each
COMPARE
Fermentation DO Probe; SS/Polarographic/150 mm, 12 mm
₹175,000.00INR / Each