Cole-Parmer Fermentation DO Probe; SS/Polarographic/320 mm, 3/4" NPT
Representative image only.
Cole-Parmer Fermentation DO Probe; SS/Polarographic/320 mm, 3/4" NPT
Cole-Parmer | Item: 0572602
Designed for fermentation applications where probes are repeatedly steam-sterilized
₹210,000.00INR / Each
₹210,000.00INR / Each
Buy It Together
Current Item
Cole-Parmer Fermentation DO Probe; SS/Polarographic/320 mm, 3/4" NPT
Cole-Parmer Fermentation DO Probe; SS/Polarographic/320 mm, 3/4" NPT
0572602
₹210,000.00INR / Each
+ =
Total Price 231000.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • 316 SS Construction
  • Polarographic
  • 1 year warranty
More About this Item
All models feature a polarographic sensor, and are available in a variety of body diameters, mounting styles, and immersion lengths. Temperature compensated probes feature 100 Ω RTD temperature measuring sensors. Order cables separately.
1 Year Warranty
Specifications
  • Meter TypeAccessory; Probes and Sensors
  • Min Temperature (° C)0
  • Max Temperature (° C)80
  • Min Temperature (° F)32
  • Max Temperature (° F)176
  • Temperature compensationAutomatic
  • Length (cm)32
  • DescriptionFermentation DO Probe; SS/Polarographic/320 mm, 3/4" NPT