Cole-Parmer® Snap Seal Vials
Versatile design provide superb performance at an economical price
  • Use with crimp or snap top closures
  • Ideal for autosamplers
The Snap Seal Vial can be securely sealed with the Poly Crimp Seal, or Snap Top Caps. More glass in the neck area minimizes breakage during decapping. The Step Vial design precisely centers a limited volume insert in the vial neck, assuring dependable, reliable use with autosamplers.
3,200.00 - ₹4,000.00INR / Pkg of 100
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (mm) Outside Diameter (mm) Volume (mL)
Availability Pricing
COMPARE
32 12 2
₹3,200.00INR / Pkg of 100
COMPARE
32 12 2
₹3,700.00INR / Pkg of 100
COMPARE
32 12 2
₹4,000.00INR / Pkg of 100