Cole-Parmer Rotisseries and Accessories for the RotoFlex and RotoFlex Plus Tube Rotators
Adapt to different racks without tools to accommodate a wide range of tubes
  • Quickly and easily switch rotisseries for enhanced versitility
  • Compatible with the RotoFlex and RotoFlex Plus systems
These rotisseries are compatible with the RotoFlex and RotoFlex Plus benchtop tube rotating shakers (sold separately). There are many paddle options to choose from based on number of tubes held as well as tube size that is accommodated.
The magnetic platform, model 04397-42, can only be used with the RotoFlex Plus system. This is ideal to optimize Western Blot hybridization. Simply attach your plastic bag containing membrane and antibody solution to the plate using the metal strips provided. These magnetic strips can be purchased separately as well as part number 04397-43.
2,200.00 - ₹42,100.00INR / Pkg of 2
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Tube Rotator Rotisserie, 0.5 to 0.8 mL Tubes; 2/PK
₹4,000.00INR / Pkg of 2
COMPARE
Tube Rotator Rotisserie, 1.5 to 2.0 mL Tubes; 2/PK
₹4,000.00INR / Pkg of 2
COMPARE
Tube Rotator Rotisserie, 5 to 7 mL Tubes; 2/PK
₹4,000.00INR / Pkg of 2
COMPARE
Tube Rotator Rotisserie, 10 to 15 mL Tubes; 2/PK
₹4,000.00INR / Pkg of 2
COMPARE
Tube Rotator Rotisserie, 50 mL Tubes; 1/Ea
₹4,000.00INR / Pkg of 1
COMPARE
RotoFlex Plus Magnetic Platform; 1/Ea
₹42,100.00INR / Each
COMPARE
RotoFlex Plus Magnetic Platform Strips; 4/Pk
₹2,200.00INR / Pkg of 4
Showing 1- 7 of 7