Cole-Parmer RotoFlex™ Plus Magnetic Platform Strips; 4/Pk
Representative image only.
Cole-Parmer RotoFlex™ Plus Magnetic Platform Strips; 4/Pk
Cole-Parmer | Item: 0439743
Adapt to different racks without tools to accommodate a wide range of tubes
₹2,200.00INR / Pkg of 4
₹2,200.00INR / Pkg of 4
Buy It Together
Current Item
Cole-Parmer RotoFlex™ Plus Magnetic Platform Strips; 4/Pk
Cole-Parmer RotoFlex™ Plus Magnetic Platform Strips; 4/Pk
0439743
₹2,200.00INR / Pkg of 4
+
Cole-Parmer RotoFlex™ Benchtop Tube Rotator; 120 VAC
0439733
₹74,300.00INR / Each
=
Total Price 76500.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Quickly and easily switch rotisseries for enhanced versitility
  • Compatible with the RotoFlex and RotoFlex Plus systems
More About this Item
These rotisseries are compatible with the RotoFlex and RotoFlex Plus benchtop tube rotating shakers (sold separately). There are many paddle options to choose from based on number of tubes held as well as tube size that is accommodated.
The magnetic platform, model 04397-42, can only be used with the RotoFlex Plus system. This is ideal to optimize Western Blot hybridization. Simply attach your plastic bag containing membrane and antibody solution to the plate using the metal strips provided. These magnetic strips can be purchased separately as well as part number 04397-43.
Specifications
  • DescriptionRotoFlex Plus Magnetic Platform Strips; 4/Pk