Cole-Parmer® RotoFlex Tube Rotator
Adapt to different racks without tools to accommodate a wide range of tubes
  • Mix micro to centrifuge tubes—0.5 mL to 50 mL tubes— with these versatile rotators
  • Adjust the mixing angle of each rotisserie platform separately from 0 to 90°
  • Rotate at a fixed speed with the RotoFlex, or from 10 to 40 rpm with the RotoFlex Plus
  • Change the mixing direction to suit your application needs
These flexible tube rotators work with tubes sizes from 0.5 to 50 mL. The 360° rotation gently and thoroughly mixes samples. Additional features include an adjustable mixing angle from 0 to 90° for each of the 4 mixing panels independently.
The RotoFlex system has a fixed speed or 22 rpm for the 120V version or 18 rpm for the 240V version, and the rotational direction is reversible by simply tapping the rotisserie in the direction required. The RotoFlex Plus system has a variable speed range of 10 to 40 rpm, with a 1 rpm resolution. The Plus system also has an oscillation mode that agitates tubes while rotating for additional sample mixing. Additional rotisseries are available for purchase separately.
73,600.00 - ₹84,100.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm) Power (VAC) Description
Availability Pricing
COMPARE
22 120 RotoFlex Benchtop Tube Rotator; 120 VAC
₹74,300.00INR / Each
COMPARE
18 240 RotoFlex Benchtop Tube Rotator; 220 VAC
₹73,700.00INR / Each
COMPARE
40 120 RotoFlex Plus Digital Benchtop Tube Rotator; 120 VAC
₹84,100.00INR / Each
COMPARE
40 240 RotoFlex Plus Digital Benchtop Tube Rotator; 220 VAC
₹73,600.00INR / Each