Cole-Parmer RotoFlex™ Tube Rotator Rotisserie, 50 mL Tubes; 1/Ea

Representative image only.

Cole-Parmer RotoFlex™ Tube Rotator Rotisserie, 50 mL Tubes; 1/Ea

Cole-Parmer | Item: 0439739

Adapt to different racks without tools to accommodate a wide range of tubes

₹4,000.00INR / Pkg of 1

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

₹4,000.00INR / Pkg of 1

Buy It Together

Current Item

Cole-Parmer RotoFlex™ Tube Rotator Rotisserie, 50 mL Tubes; 1/Ea
Cole-Parmer RotoFlex™ Tube Rotator Rotisserie, 50 mL Tubes; 1/Ea
0439739

₹4,000.00INR / Pkg of 1
+
Cole-Parmer RotoFlex™ Benchtop Tube Rotator; 120 VAC
0439733

₹74,300.00INR / Each
+ =
Total Price 78300.00 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Quickly and easily switch rotisseries for enhanced versitility
  • Compatible with the RotoFlex and RotoFlex Plus systems

More About this Item

These rotisseries are compatible with the RotoFlex and RotoFlex Plus benchtop tube rotating shakers (sold separately). There are many paddle options to choose from based on number of tubes held as well as tube size that is accommodated.
The magnetic platform, model 04397-42, can only be used with the RotoFlex Plus system. This is ideal to optimize Western Blot hybridization. Simply attach your plastic bag containing membrane and antibody solution to the plate using the metal strips provided. These magnetic strips can be purchased separately as well as part number 04397-43.

Specifications
  • Number of Tubes6
  • Tube Size28 to 29 mm OD
  • DescriptionTube Rotator Rotisserie, 50 mL Tubes; 1/Ea
Reviews
Q&A