Cole-Parmer® Replacement Ionic Strength Adjustors
Reduce measurement error
  • Replacement ionic strength adjustors come in 475-mL bottles
Ensure consistent and accurate readings from your ISE with these ionic strength adjustors.
7,000.00 - ₹29,400.00INR / Gallon of 1
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Ion-Selective Electrode Reference Electrolyte Description
Availability Pricing
COMPARE
5 M NaNO3 Replacement ionic strength adjustors, 5 M NaNO3
₹10,300.00INR / Pint of 1
COMPARE
Calcium, Water hardness 4 M KCl Replacement ionic strength adjustors, 4 M KCl
₹7,000.00INR / Pint of 1
COMPARE
Ammonium, Potassium 5 M NaCl Replacement ionic strength adjustors, 5 M NaCl
₹10,600.00INR / Pint of 1
COMPARE
Sodium 4 M NH4Cl with 4 M NH4OH Replacement ionic strength adjustor, 4M NH4Cl with 4M NH4OH; 475 mL
Call For Price
COMPARE
Fluoride TiSAB 1 Replacement ionic strength adjustor, TISAB I, 1 gallon
₹29,400.00INR / Gallon of 1
COMPARE
Fluoroborate, Lithium, Nitrate 2 M (NH4)2SO4 Replacement ionic strength adjustor, 2M (NH4)2SO4; 475 mL
₹10,600.00INR / Pint of 1