Cole-Parmer Replacement ionic strength adjustor, TISAB I, 1 gallon
Representative image only.
Cole-Parmer Replacement ionic strength adjustor, TISAB I, 1 gallon
Cole-Parmer | Item: 2750358
Reduce measurement error
₹29,400.00INR / Gallon of 1
₹29,400.00INR / Gallon of 1
Buy It Together
Current Item
Cole-Parmer Replacement ionic strength adjustor, TISAB I, 1 gallon
Cole-Parmer Replacement ionic strength adjustor, TISAB I, 1 gallon
2750358
₹29,400.00INR / Gallon of 1
+
Cole-Parmer Copper (Cu+2) ISE Double junction solution kits
2750265
₹22,300.00INR / Each
+ + =
Total Price 131500.00 ADD 4 ITEMS TO CART
Product Description
Key Features
  • Replacement ionic strength adjustors come in 475-mL bottles
  • 1 year warranty
More About this Item
Ensure consistent and accurate readings from your ISE with these ionic strength adjustors.
1 Year Warranty
Specifications
  • Reference ElectrolyteTiSAB 1
  • Ion-Selective ElectrodeFluoride
  • DescriptionReplacement ionic strength adjustor, TISAB I, 1 gallon