Cole-Parmer Replacement ionic strength adjustor, 4M NH4Cl with 4M NH4OH; 475 mL
Representative image only.
Cole-Parmer Replacement ionic strength adjustor, 4M NH4Cl with 4M NH4OH; 475 mL
Cole-Parmer | Item: 2750354
Reduce measurement error
Call For Price
Call For Price
Buy It Together
Current Item
Cole-Parmer Replacement ionic strength adjustor, 4M NH4Cl with 4M NH4OH; 475 mL
Cole-Parmer Replacement ionic strength adjustor, 4M NH4Cl with 4M NH4OH; 475 mL
2750354
Call For Price
+
Cole-Parmer Copper (Cu+2) ISE Double junction solution kits
2750265
₹22,300.00INR / Each
+ + =
Total Price 102100.00 ADD 4 ITEMS TO CART
Product Description
Key Features
  • Replacement ionic strength adjustors come in 475-mL bottles
  • 1 year warranty
More About this Item
Ensure consistent and accurate readings from your ISE with these ionic strength adjustors.
1 Year Warranty
Specifications
  • Reference Electrolyte4 M NH4Cl with 4 M NH4OH
  • Ion-Selective ElectrodeSodium
  • DescriptionReplacement ionic strength adjustor, 4M NH4Cl with 4M NH4OH; 475 mL