Cole-Parmer® Quartz Glass Crucibles
Ideal for applications that require ashing of samples!
  • Offers low expansion, dimensional stability, and thermal shock resistance
  • Withstand temperatures up to 1050  °C (1922  °F)
These clear quartz glass crucibles are ideal for your analytical applications involving sample ashing. Made from quartz, they offer low thermal expansion, excellent dimensional stability, and high resistance to thermal shock. They can withstand continuous operating temperatures up to 1050 °C (1922 °F).
Lids are also manufactured from quartz and sold separately.
7,800.00 - ₹86,500.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Top Outside Diameter (mm) Weight (g)
Availability Pricing
COMPARE
15 41 12
₹7,800.00INR / Each
COMPARE
25 47 16
₹20,900.00INR / Each
COMPARE
50 57 34
₹29,500.00INR / Each
COMPARE
80 67 35
₹59,300.00INR / Each
COMPARE
100 70 43
₹69,400.00INR / Each
COMPARE
150 82 52
₹86,500.00INR / Each