Cole-Parmer® Petri Dishes
Designed for easy one hand opening
and closing operations
  • For single use only
  • Vented for easy gas exchange and evaporation thereby
  • reduce condensation during incubation
  • MOC: Heavy Metal free Hydrophobic Polystyrene
  • Optically Clear top and bottom viewing surfaces
  • Sterilized by Gamma Irradiation with SAL 10-6
5,900.00 - ₹22,700.00INR / Case of 500
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Description
Availability Pricing
COMPARE
Polystyrene Cole-Parmer® Petri Dish, Polystyrene, 90 mm, Individually wrapped, Sterile; 500/Case
₹6,100.00INR / Case of 500
COMPARE
Polystyrene Cole-Parmer® Petri Dish, Polystyrene, 90 mm, Sterile; 10 pcs/sleeve; 56 sleeves/Case
₹5,900.00INR / Case of 560
COMPARE
Polystyrene Cole-Parmer® Petri Dish, Polystyrene, 90 mm, Double bagged, Sterile; 20 pcs/sleeve; 28 sleeves/Case
₹6,200.00INR / Case of 560
COMPARE
Polystyrene Cole-Parmer® Contact Plate, Polystyrene, 55 mm, Individually wrapped, Sterile; 1080/Case
₹12,500.00INR / Case of 1000
COMPARE
Polystyrene Cole-Parmer® Contact Plate, Polystyrene, 55 mm, Sterile; 10 pcs/sleeve; 108 sleeves/Case
₹11,900.00INR / Case of 1080
COMPARE
Polystyrene Cole-Parmer® Contact Plate, Polystyrene, 55 mm, Double bagged, Sterile; 20 pcs/sleeve; 54 sleeves/Case
₹12,200.00INR / Case of 1080
COMPARE
Polystyrene Cole-Parmer® Contact Plate, Lockable, Polystyrene, 55 mm, Sterile; 10 pcs/sleeve; 108 sleeves/Case
₹22,700.00INR / Case of 1080
Showing 1- 7 of 7